2-Hydroxy-p-toluic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-4-methylbenzoic acid, m-Homosalicyclic acid, m-Cresotic acid, 4-Methylsalicylic acid

A131260-100G A131260-25G
77246-550EA 15 USD
77246-550 77246-552 77246-554 77246-556
2-Hydroxy-p-toluic acid 97%
2-Hydroxy-p-toluic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MDL Number: MFCD00002454
CAS Number: 50-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR