(2-Aminopyridin-3-yl)methyl tert-butyl carbonate 97%

Supplier: Ambeed
Warning

A131121-100MG A131121-10G A131121-1G A131121-250MG A131121-5G
77272-000EA 8.24 USD
77272-000 77272-002 77272-004 77272-006 77272-008
(2-Aminopyridin-3-yl)methyl tert-butyl carbonate 97%
(2-Aminopyridin-3-yl)methyl tert-butyl carbonate

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₃
MW: 224.25 g/mol
MDL Number: MFCD31619103
CAS Number: 2153472-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR