2-Benzoylpyridine 99%

Supplier: Ambeed
Warning

Synonyms: Phenyl(pyridin-2-yl)methanone, Phenyl 2-pyridyl ketone

A131032-1000G A131032-100G A131032-10G A131032-25G A131032-500G A131032-5G
77258-456EA 505.73 USD
77258-456 77258-458 77258-460 77258-462 77258-464 77258-466
2-Benzoylpyridine 99%
2-Benzoylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₉NO
MW: 183.21 g/mol
MDL Number: MFCD00006300
CAS Number: 91-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR