6-Amino-1-methyluracil 97%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-1-methylpyrimidine-2,4(1H,3H)-dione

A130941-100G A130941-25G A130941-500G A130941-5G
77285-306EA 52.53 USD
77285-306 77285-308 77285-310 77285-312
6-Amino-1-methyluracil 97%
6-Amino-1-methyluracil

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂
MW: 141.13 g/mol
MDL Number: MFCD00075366
CAS Number: 2434-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR