5-Phenyl-1H-pyrazol-3(2H)-one 98%

Supplier: Ambeed
Warning

A130904-1G A130904-250MG
77234-514EA 47 USD
77234-514 77234-516
5-Phenyl-1H-pyrazol-3(2H)-one 98%
5-Phenyl-1H-pyrazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD00956881
CAS Number: 27412-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR