5,7-Dihydroxy-3-(3-hydroxy-4,5-dimethoxyphenyl)-6-methoxy-4H-chromen-4-one 97%

Supplier: Ambeed
Warning

A130806-10MG A130806-1MG A130806-25MG A130806-5MG
77246-522EA 53 USD
77246-522 77246-524 77246-526 77246-528
5,7-Dihydroxy-3-(3-hydroxy-4,5-dimethoxyphenyl)-6-methoxy-4H-chromen-4-one 97%
5,7-Dihydroxy-3-(3-hydroxy-4,5-dimethoxyphenyl)-6-methoxy-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₈H₁₆O₈
MDL Number: MFCD00597047
CAS Number: 548-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR