1-Methyl-1H-benzo[d]imidazole-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A130547-1G A130547-250MG A130547-5G
77234-494EA 95 USD
77234-494 77234-496 77234-498
1-Methyl-1H-benzo[d]imidazole-6-carboxylic acid 95%
1-Methyl-1H-benzo[d]imidazole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD11043014
CAS Number: 53484-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR