5-Bromo-2-(5-bromo-2-thienyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-(5-bromothiophen-2-yl)pyridine, 2-Bromo-5-(5-bromo-2-pyridyl)thiophene

A130532-100MG A130532-250MG A130532-50MG
77234-488EA 20.6 USD
77234-488 77234-490 77234-492
5-Bromo-2-(5-bromo-2-thienyl)pyridine 98%
5-Bromo-2-(5-bromo-2-thienyl)pyridine

Precaución: for Research Use Only

Formula: C₉H₅Br₂NS
MW: 319.01 g/mol
MDL Number: MFCD29765747
CAS Number: 136902-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR