1-Ethyl-3-methylimidazolium ethylsulphate 98%

Supplier: Ambeed
Danger

Synonyms: EMIM ESU

A130527-25G A130527-5G
77234-484EA 22 USD
77234-484 77234-486
1-Ethyl-3-methylimidazolium ethylsulphate 98%
1-Ethyl-3-methylimidazolium ethylsulphate

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₄S
MDL Number: MFCD06798189
CAS Number: 342573-75-5
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR