6-(Benzyloxy)-1H-indole 96%

Supplier: Ambeed
Warning

A130335-10G A130335-1G A130335-25G A130335-100G A130335-5G
77246-510EA 70 USD
77246-510 77246-512 77246-514 77549-544 77615-030
6-(Benzyloxy)-1H-indole 96%
6-(Benzyloxy)-1H-indole

Precaución: for Research Use Only

Formula: C15H13NO
MDL Number: MFCD00053554
CAS Number: 15903-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR