Methyl 4-(bromoacetyl)benzoate 97%

Supplier: Ambeed
Danger

Synonyms: 4-(Methoxycarbonyl)phenacyl bromide, Methyl 4-(2-bromoacetyl)benzoate, 2-Bromo-4'-(methoxycarbonyl)acetophenone

A130291-100MG A130291-1G A130291-250MG A130291-5G A130291-25G
77285-288EA 8 USD
77285-288 77285-290 77285-292 77566-532 77615-028
Methyl 4-(bromoacetyl)benzoate 97%
Methyl 4-(bromoacetyl)benzoate

Precaución: For research use only.

Formula: C₁₀H₉BrO₃
MW: 257.081 g/mol
MDL Number: MFCD08062347
CAS Number: 56893-25-5
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR