N-(3-Chloro-4-fluorophenyl)-1H-pyrazolo[4,3-b]pyridin-3-amine 97%
Supplier: Ambeed
|
Warning
|
A130260-100MG
A130260-250MG
77246-506EA
41.2
USD
77246-506
77246-508
N-(3-Chloro-4-fluorophenyl)-1H-pyrazolo[4,3-b]pyridin-3-amine 97%
N-(3-Chloro-4-fluorophenyl)-1H-pyrazolo[4,3-b]pyridin-3-amine
Precaución: for Research Use Only
Formula:
C₁₂H₈ClFN₄ MW: 262.67 g/mol |
MDL Number:
MFCD30481312 CAS Number: 1330624-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...