4-Bromoquinoline-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A130145-100MG A130145-250MG
77285-286EA 75.19 USD
77285-286 77747-931
4-Bromoquinoline-2-carboxylic acid 97%
4-Bromoquinoline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₂
MW: 252.06 g/mol
MDL Number: MFCD08705670
CAS Number: 209003-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR