5-(Isopropylamino)pentanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(Isopropylamino)pentan-1-ol
A130096-1G
A130096-250MG
A130096-5G
77234-474EA
78
USD
77234-474
77234-476
77234-478
5-(Isopropylamino)pentanol 97%
5-(Isopropylamino)pentanol
Precaución: for Research Use Only
Formula:
C₈H₁₉NO MW: 145.25 g/mol |
MDL Number:
MFCD14708174 CAS Number: 40447-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...