4-Bromo-5-chloro-2-nitroaniline 95%
Supplier: Ambeed
|
Warning
|
A130075-10G
A130075-1G
A130075-250MG
A130075-25G
A130075-5G
77285-276EA
29
USD
77285-276
77285-278
77285-280
77285-282
77285-284
4-Bromo-5-chloro-2-nitroaniline 95%
4-Bromo-5-chloro-2-nitroaniline
Precaución: for Research Use Only
Formula:
C₆H₄BrClN₂O₂ |
MDL Number:
MFCD11100159 CAS Number: 827-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...