2-Chloro-4-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A129903-100G A129903-1G A129903-5G A129903-10G
77258-418EA 334.99 USD
77258-418 77258-420 77258-422 77592-500
2-Chloro-4-methylbenzaldehyde 98%
2-Chloro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO
MDL Number: MFCD10703515
CAS Number: 50817-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR