Ethyl-1H-indazole-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1H-indazole-3-carboxylate

A129797-100MG A129797-10G A129797-1G A129797-250MG A129797-25G A129797-5G
77234-458EA 5.15 USD
77234-458 77234-460 77234-462 77234-464 77234-466 77234-468
Ethyl-1H-indazole-3-carboxylate 95%
Ethyl 1H-indazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
MDL Number: MFCD01138134
CAS Number: 4498-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR