2-Chloro-6-nitroanisole 97%

Supplier: Ambeed
Warning

A129795-1G A129795-25G A129795-5G
77258-412EA 11 USD
77258-412 77258-414 77258-416
2-Chloro-6-nitroanisole 97%
2-Chloro-6-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MDL Number: MFCD00065066
CAS Number: 80866-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR