2-(2,4,6-Trichlorophenoxy)ethanol 98%

Supplier: Ambeed
Warning

A129753-1G A129753-5G
77234-454EA 14 USD
77234-454 77234-456
2-(2,4,6-Trichlorophenoxy)ethanol 98%
2-(2,4,6-Trichlorophenoxy)ethanol

Precaución: for Research Use Only

Formula: C₈H₇Cl₃O₂
MDL Number: MFCD02231270
CAS Number: 6161-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR