2,3-Difluoro-5-(1-methyl-1H-pyrazol-4-yl)pyridine 97%
Supplier: Ambeed
|
Warning
|
A129701-1G
A129701-250MG
77258-408EA
48
USD
77258-408
77258-410
2,3-Difluoro-5-(1-methyl-1H-pyrazol-4-yl)pyridine 97%
2,3-Difluoro-5-(1-methyl-1H-pyrazol-4-yl)pyridine
Precaución: for Research Use Only
Formula:
C₉H₇F₂N₃ |
MDL Number:
MFCD28366017 CAS Number: 1151801-90-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...