4-(2-Chloropyrimidin-4-yl)morpholine 97%

Supplier: Ambeed
Warning

A129686-1G A129686-250MG
77285-258EA 74.16 USD
77285-258 77285-260
4-(2-Chloropyrimidin-4-yl)morpholine 97%
4-(2-Chloropyrimidin-4-yl)morpholine

Precaución: for Research Use Only

Formula: C₈H₁₀ClN₃O
MDL Number: MFCD00981910
CAS Number: 62968-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR