AZD-5153 98+%

Supplier: Ambeed
Danger

Synonyms: 6-hydroxynaphthalene-2-carboxylic acid, 4-[2-[4-[1-(3-methoxy-[1,2,4]triazolo[4,3-b]pyridazin-6-yl)piperidin-4-yl]phenoxy]ethyl]-1,3-dimethylpiperazin-2-one, AZD5153 6-Hydroxy-2-naphthoic acid, (R)-4-(2-(4-(1-(3-Methoxy-[1,2,4]triazolo[4,3-b]pyridazin-6-yl)piperidin-4-yl)phenoxy)ethyl)-1,3-dimethylpiperazin-2-one 6-hydroxy-2-naphthoate

A129684-10MG A129684-1MG A129684-25MG A129684-50MG A129684-5MG
77271-938EA 185 USD
77271-938 77271-940 77271-942 77271-944 77271-946
AZD-5153 98+%
AZD-5153

Precaución: for Research Use Only

Formula: C₂₅H₃₃N₇O₃∙C₁₁H₈O₃
MDL Number: MFCD31382122
CAS Number: 1869912-40-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR