Ethyl-2,4-dimethylpyrrole-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2,4-dimethyl-1H-pyrrole-3-carboxylate

A129598-100G A129598-10G A129598-1G A129598-250MG A129598-25G A129598-500G A129598-5G
77246-476EA 366.68 USD
77246-476 77246-478 77246-480 77246-482 77246-484 77246-486 77246-488
Ethyl-2,4-dimethylpyrrole-3-carboxylate 97%
Ethyl 2,4-dimethylpyrrole-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MW: 167.2050 g/mol
MDL Number: MFCD01365811
CAS Number: 2199-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR