4-Hydroxyphenylretinamide 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Hydroxyphenyl)-all- trans retinamide, Retinoic Acid p-Hydroxyanilide, (2E,4E,6E,8E)-N-(4-Hydroxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenamide, all-trans-N-(4-Hydroxyphenyl)retinamide, Fenretinide, 4-HPR

A129384-10MG A129384-1MG A129384-50MG
77285-246EA 43.26 USD
77285-246 77285-248 77285-250
4-Hydroxyphenylretinamide 98%
4-Hydroxyphenylretinamide

Precaución: for Research Use Only

Formula: C₂₆H₃₃NO₂
MW: 391.56 g/mol
MDL Number: MFCD00792674
CAS Number: 65646-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR