Boc-L-leucine monohydrate 95%

Supplier: Ambeed
Warning

Synonyms: Boc-Leu-OH·H₂O, Boc-Leu-OH·H2O, N-(tert-Butoxycarbonyl)-L-leucine monohydrate

A129371-1000G A129371-100G A129371-25G
77271-922EA 107 USD
77271-922 77271-924 77271-926 77271-928
Boc-L-leucine monohydrate 95%
Boc-L-leucine monohydrate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄·H₂O
MW: 249.307 g/mol
MDL Number: MFCD00065582
CAS Number: 200936-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR