D-4,4'-Biphenylalanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Phenyl-D-phenyalanine, H-4-Phenyl-D-Phe-OH, 3-(4-Biphenylyl)-D-alanine, (R)-3-([1,1'-Biphenyl]-4-yl)-2-aminopropanoic acid
A129302-1G
A129302-250MG
A129302-25G
A129302-5G
77258-404EA
33
USD
77258-404
77258-406
77747-923
77747-924
D-4,4'-Biphenylalanine 97%
D-4,4'-Biphenylalanine
Precaución: for Research Use Only
Formula:
C₁₅H₁₅NO₂ MW: 241.29 g/mol |
MDL Number:
MFCD01318879 CAS Number: 170080-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...