3-Amino-4-methoxybenzamide 99%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-p-anisamide

A129186-25G A129186-5G
77271-910EA 59 USD
77271-910 77271-912
3-Amino-4-methoxybenzamide 99%
3-Amino-4-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD00017132
CAS Number: 17481-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR