2-Chloroquinazolin-4-amine 96%

Supplier: Ambeed
Warning

A129168-100MG A129168-1G A129168-250MG A129168-5G
77258-396EA 7.21 USD
77258-396 77258-398 77258-400 77258-402
2-Chloroquinazolin-4-amine 96%
2-Chloroquinazolin-4-amine

Precaución: for Research Use Only

Formula: C₈H₆ClN₃
MW: 179.60 g/mol
MDL Number: MFCD08234547
CAS Number: 59870-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR