4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 3-iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine

A129157-100G A129157-100MG A129157-10G A129157-1G A129157-250MG A129157-25G A129157-5G A129157-500G
77285-228EA 241.02 USD
77285-228 77285-230 77285-232 77285-234 77285-236 77285-238 77285-240 77747-922
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine 98%
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₅H₄N₅I
MW: 261.03 g/mol
MDL Number: MFCD03787931
CAS Number: 151266-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR