(3AS,3bR,7aS,8aS)-2,2,5,5-tetramethyltetrahydro-3aH-[1,3]dioxolo[4',5':4,5]furo[3,2-d][1,3]dioxine 97%

Supplier: Ambeed
Warning

A129152-100MG A129152-1G A129152-250MG
77258-390EA 50 USD
77258-390 77258-392 77258-394
(3AS,3bR,7aS,8aS)-2,2,5,5-tetramethyltetrahydro-3aH-[1,3]dioxolo[4',5':4,5]furo[3,2-d][1,3]dioxine 97%
(3AS,3bR,7aS,8aS)-2,2,5,5-tetramethyltetrahydro-3aH-[1,3]dioxolo[4',5':4,5]furo[3,2-d][1,3]dioxine

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₅
MW: 230.26 g/mol
MDL Number: MFCD00004946
CAS Number: 131156-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR