2,4-Diaminoquinazoline 98%

Supplier: Ambeed
Warning

A129144-1G A129144-250MG A129144-5G
77246-452EA 12 USD
77246-452 77246-454 77246-456
2,4-Diaminoquinazoline 98%
2,4-Diaminoquinazoline

Precaución: for Research Use Only

Formula: C₈H₈N₄
MDL Number: MFCD00236032
CAS Number: 1899-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR