2,4,5-Trifluoro-3-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A129143-1000G A129143-100G A129143-10G A129143-25G A129143-500G A129143-5G
77234-420EA 192 USD
77234-420 77234-422 77234-424 77234-426 77234-428 77745-933
2,4,5-Trifluoro-3-methoxybenzoic acid 98%
2,4,5-Trifluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₃
MDL Number: MFCD00153201
CAS Number: 112811-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR