(3-Nitrophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A129140-100G A129140-10G A129140-1G A129140-250MG A129140-25G A129140-5G
77234-408EA 257.5 USD
77234-408 77234-410 77234-412 77234-414 77234-416 77234-418
(3-Nitrophenyl)acetic acid 97%
(3-Nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄(NO₂))CH₂COOH
MW: 181.15 g/mol
MDL Number: MFCD00007278
CAS Number: 1877-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR