6'-((6-Aminopyrimidin-4-yl)amino)-8'-methyl-1'H-spiro[cyclohexane-1,3'-imidazo[1,5-a]pyridine]-1',5'(2'H)-dione 98%

Supplier: Ambeed
Warning

Synonyms: Tomivosertib

A129059-5MG A129059-10MG A129059-1MG A129059-50MG
77258-388EA 30.9 USD
77258-388 77747-919 77747-920 77747-921
6'-((6-Aminopyrimidin-4-yl)amino)-8'-methyl-1'H-spiro[cyclohexane-1,3'-imidazo[1,5-a]pyridine]-1',5'(2'H)-dione 98%
6'-((6-Aminopyrimidin-4-yl)amino)-8'-methyl-1'H-spiro[cyclohexane-1,3'-imidazo[1,5-a]pyridine]-1',5'(2'H)-dione

Precaución: for Research Use Only

Formula: C₁₇H₂₀N₆O₂
MW: 340.38 g/mol
MDL Number: MFCD30489732
CAS Number: 1849590-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR