(2,3,4-Trifluorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A128887-1G A128887-250MG A128887-25G
77234-378EA 17 USD
77234-378 77234-380 77745-929
(2,3,4-Trifluorophenyl)acetic acid 97%
(2,3,4-Trifluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₂F₃CH₂COOH
MDL Number: MFCD00061235
CAS Number: 243666-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR