2',5'-Dimethoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,5-Dimethoxyphenyl)ethan-1-one, 1-(2,5-Dimethoxyphenyl)ethanone

A128794-100G A128794-25G A128794-5G
77246-438EA 39.14 USD
77246-438 77246-440 77745-928
2',5'-Dimethoxyacetophenone 98%
2',5'-Dimethoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD00008728
CAS Number: 1201-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR