1-Boc-4-(4-nitrophenyl)piperazine 98%

Supplier: Ambeed
Warning

A128710-1G A128710-5G
77285-200EA 13 USD
77285-200 77285-202
1-Boc-4-(4-nitrophenyl)piperazine 98%
1-Boc-4-(4-nitrophenyl)piperazine

Precaución: for Research Use Only

Formula: C₁₅H₂₁N₃O₄
MDL Number: MFCD02187326
CAS Number: 182618-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR