3-(Ethoxydimethylsilyl)propyl methacrylate 95+%

Supplier: Ambeed
Warning

Synonyms: Methacrylic acid 3-(Ethoxydimethylsilyl)propyl ester, (3-Methacryloyloxy)propyldimethylethoxysilane

A128650-1G A128650-25G A128650-5G
77271-898EA 8.24 USD
77271-898 77271-900 77271-902
3-(Ethoxydimethylsilyl)propyl methacrylate 95+%
3-(Ethoxydimethylsilyl)propyl methacrylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂O₃Si
MW: 230.38 g/mol
MDL Number: MFCD00053877
CAS Number: 13731-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR