1-(Tetrahydro-2H-pyran-2-yl)-5-(trifluoromethyl)-1H-indazol-4-ol 95%

Supplier: Ambeed
Warning

A128516-100MG A128516-1G A128516-250MG
77271-892EA 91.67 USD
77271-892 77271-894 77271-896
1-(Tetrahydro-2H-pyran-2-yl)-5-(trifluoromethyl)-1H-indazol-4-ol 95%
1-(Tetrahydro-2H-pyran-2-yl)-5-(trifluoromethyl)-1H-indazol-4-ol

Precaución: for Research Use Only

Formula: C₁₃H₁₃F₃N₂O₂
MW: 286.25 g/mol
MDL Number: MFCD31619101
CAS Number: 2197061-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR