3-Hydroxy-2-iodo-6-methylpyridine 98%

Supplier: Ambeed
Warning

A128485-100MG A128485-1G A128485-250MG A128485-5G A128485-10G
77258-356EA 5 USD
77258-356 77258-358 77258-360 77258-362 77747-917
3-Hydroxy-2-iodo-6-methylpyridine 98%
3-Hydroxy-2-iodo-6-methylpyridine

Precaución: for Research Use Only

Formula: C6H6INO
MDL Number: MFCD00006246
CAS Number: 23003-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR