11-Methyldodecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate 97%

Supplier: Ambeed
Warning

A128364-10G A128364-1G A128364-25G A128364-5G
77258-348EA 39.14 USD
77258-348 77258-350 77258-352 77258-354
11-Methyldodecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate 97%
11-Methyldodecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate

Precaución: for Research Use Only

Formula: C₃₀H₅₂O₃
MW: 460.73 g/mol
MDL Number: MFCD31629688
CAS Number: 2206682-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR