3-Hydroxybenzophenone 98%

Supplier: Ambeed
Warning

A128305-10G A128305-1G A128305-250MG A128305-25G
77271-880EA 59 USD
77271-880 77271-882 77271-884 77271-886
3-Hydroxybenzophenone 98%
3-Hydroxybenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₂
MDL Number: MFCD00002297
CAS Number: 13020-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR