5-Bromo-2,3-diphenylpyrazine 97%

Supplier: Ambeed
Warning

A128302-10G A128302-1G A128302-250MG
77246-422EA 208 USD
77246-422 77246-424 77246-426
5-Bromo-2,3-diphenylpyrazine 97%
5-Bromo-2,3-diphenylpyrazine

Precaución: for Research Use Only

Formula: C₁₆H₁₁BrN₂
MW: 311.18 g/mol
MDL Number: MFCD00234886
CAS Number: 243472-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR