3-Phenoxybenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Hydroxymethyl)diphenyl ether, (3-Phenoxyphenyl)methanol

A128166-1000G A128166-100G A128166-10G A128166-25G A128166-500G A128166-5G
77285-178EA 224.54 USD
77285-178 77285-180 77285-182 77285-184 77285-186 77285-188
3-Phenoxybenzyl alcohol 98%
3-Phenoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₂
MW: 200.24 g/mol
MDL Number: MFCD00004636
CAS Number: 13826-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR