2,3-Dihydro[1,4]dioxino[2,3-b]pyridine 98%
Supplier: Ambeed
|
Warning
|
A128061-100MG
A128061-250MG
A128061-5G
77246-418EA
14
USD
77246-418
77246-420
77745-925
2,3-Dihydro[1,4]dioxino[2,3-b]pyridine 98%
2,3-Dihydro[1,4]dioxino[2,3-b]pyridine
Precaución: for Research Use Only
Formula:
C₇H₇NO₂ |
MDL Number:
MFCD09743547 CAS Number: 129421-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...