BAY 41-4109 Racemate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-(2-chloro-4-fluorophenyl)-2-(3,5-difluoropyridin-2-yl)-6-methyl-1,4-dihydropyrimidine-5-carboxylate
A127980-10MG
A127980-1MG
A127980-25MG
A127980-50MG
A127980-5MG
77258-338EA
198.79
USD
77258-338
77258-340
77258-342
77258-344
77258-346
BAY 41-4109 Racemate 98%
BAY 41-4109 Racemate
Precaución: for Research Use Only
Formula:
C₁₈H₁₃ClF₃N₃O₂ MW: 395.76 g/mol |
MDL Number:
MFCD30729326 CAS Number: 298708-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...