2,7-Dibromo-9,9-diphenylfluorene 97%

Supplier: Ambeed
Warning

A127868-100G A127868-10G A127868-25G A127868-5G
77246-410EA 262 USD
77246-410 77246-412 77246-414 77246-416
2,7-Dibromo-9,9-diphenylfluorene 97%
2,7-Dibromo-9,9-diphenylfluorene

Precaución: for Research Use Only

Formula: C₂₅H₁₆Br₂
MDL Number: MFCD09907958
CAS Number: 186259-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR