6-Bromo-1H-indol-4-amine 95%

Supplier: Ambeed
Warning

A127841-100MG A127841-250MG
77285-166EA 63.86 USD
77285-166 77285-168
6-Bromo-1H-indol-4-amine 95%
6-Bromo-1H-indol-4-amine

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.05 g/mol
MDL Number: MFCD03095013
CAS Number: 350800-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR