(3-Hydroxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxyphenylacetic acid

A127796-100G A127796-1G A127796-25G A127796-500G A127796-5G
77246-400EA 130.81 USD
77246-400 77246-402 77246-404 77246-406 77246-408
(3-Hydroxyphenyl)acetic acid 97%
(3-Hydroxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄(OH))CH₂COOH
MW: 152.15 g/mol
MDL Number: MFCD00004337
CAS Number: 621-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR