2,3,6-Trichloropyridine 96%

Supplier: Ambeed
Warning

A127654-100G A127654-10G A127654-25G A127654-5G
77246-382EA 125.66 USD
77246-382 77246-384 77246-386 77246-388
2,3,6-Trichloropyridine 96%
2,3,6-Trichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₃N
MW: 182.43 g/mol
MDL Number: MFCD00463516
CAS Number: 6515-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR